Novus Biologicals
Manufacturer Code:NBP182268
Catalog # NBP182268
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LVSEVDRVMEEFNLTTFYQDPSFHLSLAWCVGDARLQLEGQCLQELQAIVDGFEDAEVLLRVHTEQVRCKSGNKFFSM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf57 chromosome 16 open reading frame 57 FLJ13154 HVSL motif containing 1 HVSL1 hypothetical protein LOC79650 PN; hUsb1; HVSL motif containing 1; mutated in poikiloderma with neutropenia protein 1; putative U6 snRNA phosphodiesterase; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA phosphodiesterase; UPF0406 protein C16orf57; USB1
Gene Aliases: C16orf57; hUsb1; HVSL1; Mpn1; PN; USB1
UniProt ID: (Human) Q9H8X8
Entrez Gene ID: (Human) 79650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.