Novus Biologicals
Manufacturer Code:NBP289429AF700
Catalog # NB107567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 28 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human HVEM/TNFRSF14 (NP_003811.2 Met1-Val202). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: CD270; CD270 antigen CD40-like protein Herpes virus entry mediator A Herpesvirus entry mediator A HveA HVEMTR2HVEAATAR LIGHTR tumor necrosis factor receptor superfamily member 14 tumor necrosis factor receptor superfamily member 14 (herpesvirus entrymediator) Tumor necrosis factor receptor-like 2 tumor necrosis factor receptor-like gene2; CD40-like protein; Herpes virus entry mediator A; Herpesvirus entry mediator A; TR2; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2
Gene Aliases: ATAR; CD270; HVEA; HVEM; LIGHTR; TNFRSF14; TR2; UNQ329/PRO509
UniProt ID: (Human) Q92956
Entrez Gene ID: (Human) 8764
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.