Novus Biologicals
Manufacturer Code:NBP304397100UL
Catalog # NB095913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human HVCN1 (NP_115745.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HV1; Hydrogen voltage-gated channel 1; hydrogen voltage-gated channel 1 VSOP; Voltage sensor domain-only protein; Voltage-gated hydrogen channel 1
Gene Aliases: HV1; HVCN1; UNQ578/PRO1140; VSOP
UniProt ID: (Human) Q96D96
Entrez Gene ID: (Human) 84329
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.