Invitrogen
Manufacturer Code:PA520374
Catalog # PIPA520374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide from near the carboxy terminus of human ARF-BP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARF-binding protein 1; ARF-BP1; BJ-HCC-24 tumor antigen; E3 ubiquitin-protein ligase HUWE1; E3Histone; HECT domain protein LASU1; HECT, UBA and WWE domain-containing protein 1; HECT-type E3 ubiquitin transferase HUWE1; HectH9; Homologous to E6AP carboxyl terminus homologous protein 9; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; RP3-339A18.4 ARF-BP1 HECTH9 HSPC272 Ib772 LASU1 MULE UREB1 ARF binding protein 1 ARF-binding protein 1 BJ-HCC-24 tumor antigen E3 ubiquitin-protein ligase HUWE1 HECT domain protein LASU1 HECT UBA and WWE domain-containing protein 1 Mcl-1 ubiquitin ligase E3 URE-B1 URE-binding protein 1 homologous to E6AP carboxyl terminus homologous protein 9 large structure of UREB1 upstream regulatory element-binding protein 1; upstream regulatory element binding protein 1; Upstream regulatory element-binding protein 1; URE-B1; URE-binding protein 1
Gene Aliases: 5430439H10Rik; ARF-BP1; AU041296; C430014N20Rik; C80292; Gm1718; HECTH9; HSPC272; HUWE1; Ib772; KIAA0312; KIAA1578; LASU1; MULE; URE-B1; UREB1
UniProt ID: (Human) Q7Z6Z7, (Mouse) Q7TMY8
Entrez Gene ID: (Human) 10075, (Rat) 501546, (Mouse) 59026
Molecular Function:
ligase
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.