Abnova
Manufacturer Code:PAB12996
Catalog # 89-116-251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 19 amino acids of human HUWE1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HUWE1.
For Research Use Only
Protein Aliases: ARF-binding protein 1; ARF-BP1; BJ-HCC-24 tumor antigen; E3 ubiquitin-protein ligase HUWE1; HECT domain protein LASU1; HECT, UBA and WWE domain-containing protein 1; HECT-type E3 ubiquitin transferase HUWE1; HECTH9; Homologous to E6AP carboxyl terminus homologous protein 9; HSPC272; Ib772; KIAA0312; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase E3; MULE; Upstream regulatory element-binding protein 1; URE-B1; URE-binding protein 1; UREB1
Gene Aliases: ARF-BP1; HECTH9; HSPC272; HUWE1; Ib772; KIAA0312; KIAA1578; LASU1; MULE; URE-B1; UREB1
UniProt ID: (Human) Q7Z6Z7
Entrez Gene ID: (Human) 10075
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.