Proteintech
Manufacturer Code:194301AP150UL
Catalog # 50-173-2403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HUWE1 Fusion Protein Ag13763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARF binding protein 1 ARF BP1 HECTH9 HSPC272 HUWE1 Ib772 KIAA0312 KIAA1578 Large structure of UREB1 LASU1 Mcl 1 ubiquitin ligase E3 MULE URE B1 URE binding protein 1 UREB1; ARF-binding protein 1; ARF-BP1; BJ-HCC-24 tumor antigen; E3 ubiquitin-protein ligase HUWE1; E3Histone; HECT domain protein LASU1; HECT, UBA and WWE domain-containing protein 1; HECT-type E3 ubiquitin transferase HUWE1; HectH9; Homologous to E6AP carboxyl terminus homologous protein 9; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; upstream regulatory element binding protein 1; Upstream regulatory element-binding protein 1; URE-B1; URE-binding protein 1
Gene Aliases: 5430439H10Rik; ARF-BP1; AU041296; C430014N20Rik; C80292; Gm1718; HECTH9; HSPC272; HUWE1; Ib772; KIAA0312; KIAA1578; LASU1; MULE; URE-B1; UREB1
UniProt ID: (Human) Q7Z6Z7, (Mouse) Q7TMY8
Entrez Gene ID: (Human) 10075, (Rat) 501546, (Mouse) 59026
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.