Proteintech
Manufacturer Code:182291AP150UL
Catalog # 50-173-2060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HUS1B Fusion Protein Ag12712 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Checkpoint protein HUS1B; Checkpoint protein HUS1B hHUS1B HUS1B RP11 532F6.1; hHUS1B; HUS1 checkpoint homolog b
Gene Aliases: HUS1B
UniProt ID: (Human) Q8NHY5
Entrez Gene ID: (Human) 135458, (Rat) 691382
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.