Invitrogen
Manufacturer Code:PA5109839
Catalog # PIPA5109839
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HUS1 (NP_004498.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Checkpoint protein HUS1; Checkpoint protein HUS1 hHUS1 Hus1 HUS1 protein mHUS1; hHUS1; HUS1 checkpoint homolog; Hus1 homolog; hus1+-like protein; mHUS1
Gene Aliases: hHUS1; HUS1
UniProt ID: (Human) O60921, (Mouse) Q8BQY8
Entrez Gene ID: (Human) 3364, (Rat) 498411, (Mouse) 15574
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.