Novus Biologicals
Manufacturer Code:NBP133589
Catalog # NBP133589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human HUS1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Checkpoint protein HUS1; checkpoint protein HUS1 hHUS1 HUS1 (S. pombe) checkpoint homolog HUS1 checkpoint homolog (S. pombe) hus1+-like protein; hHUS1; HUS1 checkpoint homolog; hus1+-like protein
Gene Aliases: hHUS1; HUS1
UniProt ID: (Human) O60921
Entrez Gene ID: (Human) 3364
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.