Proteintech
Manufacturer Code:157751AP150UL
Catalog # 50-173-0469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HTRA2 Fusion Protein Ag8455 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High temperature requirement protein A2; HtrA serine peptidase 2 HTRA2 OMI PARK13 PRSS25 Serine protease 25 Serine proteinase OMI; HtrA-like serine protease; HtrA2; motor neuron degeneration 2; Omi stress-regulated endoprotease; protease, serine, 25; Serine protease 25; Serine protease HTRA2, mitochondrial; serine protease OMI; Serine proteinase OMI
Gene Aliases: AI481710; HTRA2; mnd2; OMI; PARK13; PRSS25
UniProt ID: (Human) O43464, (Mouse) Q9JIY5
Entrez Gene ID: (Human) 27429, (Rat) 297376, (Mouse) 64704
Molecular Function:
chaperone
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.