Invitrogen
Manufacturer Code:MA537958
Catalog # PIMA537958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2238 |
Immunogen | A synthesized peptide derived from human 5HT7 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 5-HT receptor 7a 5Ht7 5-HT7 5-HT-7 5HT7 Receptor 5-HT7 receptor 5-HT-X 5-hydroxytryptamine (serotonin) receptor 7 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled) 5-hydroxytryptamine (serotonin) receptor 7 adenylate cyclase-coupled 5-hydroxytryptamine receptor 7 GPRFO high affinity serotonin receptor (5HT7) HT7 HTR7 OTTHUMP00000020077 OTTHUMP00000020078 OTTHUMP00000020079 Serotonin receptor 7; 5-HT-7; 5-HT-X; 5-HT7; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine receptor 7; GPRFO; high affinity serotonin receptor (5HT7); Serotonin receptor 7
Gene Aliases: 5-HT7; 5Ht7; HTR7
UniProt ID: (Human) P34969, (Mouse) P32304, (Rat) P32305
Entrez Gene ID: (Human) 3363, (Mouse) 15566, (Rat) 65032
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.