Invitrogen
Manufacturer Code:PA578391
Catalog # PIPA578391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of mouse 5-HT4 receptor. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5 hydroxytryptamine (serotonin) receptor 4 5HT4 5HTR4 5HT<4L> Htr4; 5-HT-4; 5-hydroxytryptamine receptor 4; Serotonin receptor 4
Gene Aliases: 5-HT-4; 5-HT4; 5-HT<4L>; 5HTR4; Htr4
UniProt ID: (Mouse) P97288
Entrez Gene ID: (Mouse) 15562
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.