Invitrogen
Manufacturer Code:PA575300
Catalog # PIPA575300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 14-66 of Human HTR4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5 hydroxytryptamine ( serotonin) receptor 4; 5-HT-4; 5-HT4 receptor (two splicing variants L and S); 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; 5-hydroxytryptamine receptor 4 5-HT-4 5-HT4 Serotonin receptor 4 HTR4; cardiac 5-HT4 receptor; Serotonin receptor 4
Gene Aliases: 5-HT-4; 5-HT4; 5-HT4R; 5-HT<4L>; 5HTR4; HTR4
UniProt ID: (Human) Q13639, (Rat) Q62758, (Mouse) P97288
Entrez Gene ID: (Human) 3360, (Rat) 25324, (Mouse) 15562
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.