Invitrogen
Manufacturer Code:PA533310
Catalog # PIPA533310
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human 5HT4 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; HTR4 5-HT4 5-HT4R 5-HT-4 5HT4 receptor 5-HT4 receptor 5-hydroxytryptamine 4 receptor 5-hydroxytryptamine receptor 4 Serotonin 5-HT-4a receptor Serotonin receptor 4 Cardiac 5-HT4 receptor Serotonin 4 receptor Serotonin 5-HT-4 receptor; Serotonin receptor 4
Gene Aliases: 5-HT4; 5-HT4R; HTR4
UniProt ID: (Human) Q13639
Entrez Gene ID: (Human) 3360
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.