Invitrogen
Manufacturer Code:PA5113610
Catalog # PIPA5113610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human HTR3C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT3-C; 5-HT3C; 5-HT3C 5-HT3-C 5-hydroxytryptamine (serotonin) receptor 3 family member C 5-hydroxytryptamine (serotonin) receptor 3C ionotropic 5-hydroxytryptamine receptor 3 subunit C 5-hydroxytryptamine receptor 3C HTR3C Serotonin receptor 3C; 5-hydroxytryptamine (serotonin) receptor 3, family member C; 5-hydroxytryptamine (serotonin) receptor 3C, ionotropic; 5-hydroxytryptamine receptor 3 subunit C; 5-hydroxytryptamine receptor 3C; Serotonin receptor 3C
Gene Aliases: HTR3C
UniProt ID: (Human) Q8WXA8
Entrez Gene ID: (Human) 170572
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.