Invitrogen
Manufacturer Code:PA5112579
Catalog # PIPA5112579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 5-hydroxytryptamine receptor 3B protein (22-238AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT3 receptor subunit B; 5-HT3 receptor subunit B 5-HT3B 5-HT-3B 5-HT3-B 5-hydroxytryptamine (serotonin) receptor 3B 5-hydroxytryptamine (serotonin) receptor 3B ionotropic 5-hydroxytryptamine 3 receptor B subunit 5-hydroxytryptamine receptor 3B HTR3B LOC100008789 Serotonin receptor 3B serotonin-gated ion channel subunit; 5-HT3-B; 5-hydroxytryptamine (serotonin) receptor 3B, ionotropic; 5-hydroxytryptamine 3 receptor B subunit; 5-hydroxytryptamine receptor 3B; Serotonin receptor 3B; serotonin-gated ion channel subunit
Gene Aliases: 5-HT3B; HTR3B
UniProt ID: (Human) O95264, (Mouse) Q9JHJ5
Entrez Gene ID: (Human) 9177, (Mouse) 57014
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.