Invitrogen
Manufacturer Code:PA577746
Catalog # PIPA577746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RPVPDWLRHLVLDR corresponding to amino acid residues 342-355 of rat 5-Hydroxytryptamine Receptor 3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-3; 5-HT3 receptor; 5-HT3-A; 5-HT3A; 5-HT3A Serotonin receptor 3A; 5-HT3R; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; 5HT3 serotonin receptor; Serotonin receptor 3A; Serotonin-gated ion channel receptor
Gene Aliases: 5-HT3; 5-HT3A; 5HT3; Htr3; Htr3a
UniProt ID: (Rat) P35563, (Mouse) P23979
Entrez Gene ID: (Rat) 79246, (Mouse) 15561
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.