Invitrogen
Manufacturer Code:PA518547
Catalog # PIPA518547
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GPQDFEKSPRDR) corresponding to the internal amino acids of HTR3A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-3; 5-HT3 receptor; 5-HT3-A; 5-HT3R; 5-hydroxytryptamine (serotonin) receptor 3A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; 5HT3 serotonin receptor; HTR3A serotonin receptor 5-hydroxytryptamine (serotonin) receptor 3A 5-HT-3 5-HT3A 5-HT3R 5HT3R HTR3 5-hydroxytryptamine (serotonin) receptor-3 5HT3 serotonin receptor Serotonin-gated ion channel receptor truncated receptor containing only 3 transmembrane domains; Serotonin receptor 3A; Serotonin-gated ion channel receptor
Gene Aliases: 5-HT-3; 5-HT3; 5-HT3A; 5-HT3R; 5HT3; 5HT3R; HTR3; HTR3A
UniProt ID: (Human) P46098, (Rat) P35563
Entrez Gene ID: (Human) 3359, (Rat) 79246
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.