Invitrogen
Manufacturer Code:PA533306
Catalog # PIPA533306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human 5HT2C Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1C; 5-HT-2C; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; HTR2C 5-HT-1C 5-HT1c receptor 5HT-1C 5-HT2C 5-HT2c receptor 5HT2C Receptor 5-HT-2C 5-HT1C 5-HTR2C HTR1C Serotonin 5-HT-1c receptor Serotonin receptor 2C Serotonin 1c receptor Serotonin 5-HT-2C receptor Serotonin 2c receptor; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; Serotonin receptor 2C
Gene Aliases: 5-HT1C; 5-HT2C; 5-HTR2C; 5HTR2C; HTR1C; HTR2C
UniProt ID: (Human) P28335
Entrez Gene ID: (Human) 3358
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.