Invitrogen
Manufacturer Code:720264
Catalog # PI720264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Protein corresponding to Human HTR2C (aa 236-311) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1C; 5-HT-2C; 5-HT1C; 5-HT2C receptor; 5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled; 5-hydroxytryptamine receptor 1C; 5-hydroxytryptamine receptor 2C; serotonin 5-HT-1C receptor; serotonin 5-HT-2C receptor; Serotonin receptor 2C; Serotonin receptor 2C 5-HT1C
Gene Aliases: 5-HT1C; 5-HT2C; 5-HT2cR; 5-HTR2C; 5HT1c; 5HTR2C; HTR1C; HTR2C; SR1
UniProt ID: (Human) P28335, (Mouse) P34968
Entrez Gene ID: (Human) 3358, (Mouse) 15560
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.