Invitrogen
Manufacturer Code:PA577749
Catalog # PIPA577749
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KMSEQ STISEHILQK corresponding to amino acid residues 6-20 of rat 5-hydroxytryptamine receptor 2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT 2B receptor; 5-HT-2B; 5-HT-2F; 5-HT2B; 5-HT2B 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; NP75 protein; Serotonin receptor 2B; Stomach fundus serotonin receptor
Gene Aliases: 5-HT(2B); 5-HT-2B; 5-HT2B; AJ012488; AV377389; HTR2B; Srl
UniProt ID: (Human) P41595, (Mouse) Q02152, (Rat) P30994
Entrez Gene ID: (Human) 3357, (Mouse) 15559, (Rat) 29581
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.