Invitrogen
Manufacturer Code:PA132494
Catalog # PIPA132494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide KLH conjugated. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT 2B receptor; 5-HT(2B) 5-HT2B; 5-HT-2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; Serotonin receptor 2B
Gene Aliases: 5-HT(2B); 5-HT-2B; 5-HT2B; HTR2B
UniProt ID: (Human) P41595
Entrez Gene ID: (Human) 3357
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.