Abnova
Manufacturer Code:PAB25513
Catalog # 89-127-126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 20 amino acids at N-terminal extracellular domain of human HTR2B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HTR2B.
For Research Use Only
Protein Aliases: 5-HT 2B receptor; 5-HT(2B); 5-HT-2B; 5-HT2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; Serotonin receptor 2B
Gene Aliases: 5-HT(2B); 5-HT-2B; 5-HT2B; HTR2B
UniProt ID: (Human) P41595
Entrez Gene ID: (Human) 3357
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.