Invitrogen
Manufacturer Code:PA595288
Catalog # PIPA595288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human 5HT2A Receptor different from the related mouse sequence by three amino acids and from the related rat sequence by two amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-2; 5-HT-2A; 5-HT2 receptor; 5-HT2A receptor; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine 2 receptor; 5-hydroxytryptamine receptor 2A; 5-hydroxytryptamine receptor 2A 5-HT-2 5-HT-2A Serotonin receptor 2A HTR2A HTR2; serotonin 5-HT-2A receptor; serotonin 5HT-2 receptor; Serotonin receptor 2A
Gene Aliases: 5-HT2A; 5Ht-2; E030013E04; Htr-2; HTR2; HTR2A
UniProt ID: (Human) P28223, (Mouse) P35363, (Rat) P14842
Entrez Gene ID: (Human) 3356, (Mouse) 15558, (Rat) 29595
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.