Invitrogen
Manufacturer Code:PA532865
Catalog # PIPA532865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from N-Terminus of human 5HT2A Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-2; 5-HT2 receptor; 5-HT2A HTR2 5-HT-2A 5-HT2 receptor 5-hydroxytryptamine receptor 2A serotonin 5-HT-2A receptor; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine receptor 2A; serotonin 5-HT-2A receptor; Serotonin receptor 2A
Gene Aliases: 5-HT2A; HTR2; HTR2A
UniProt ID: (Human) P28223
Entrez Gene ID: (Human) 3356
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.