Invitrogen
Manufacturer Code:PA5103377
Catalog # PIPA5103377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human 5-HT-2A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-2; 5-HT-2 5Ht-2 5-HT2 receptor 5-HT2A 5-HT-2A 5HT2A Receptor 5-HT2A receptor 5-HT2A/2C 5-HT2A/2C receptor 5-hydroxytryptamine (serotonin) receptor 2A 5-hydroxytryptamine (serotonin) receptor 2A G protein-coupled 5-hydroxytryptamine 2 receptor 5-hydroxytryptamine receptor 2A E030013E04 Htr2 Htr-2 HTR2A serotonin 5HT-2 receptor serotonin 5-HT-2A receptor Serotonin receptor 2A SR2A; 5-HT-2A; 5-HT2 receptor; 5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled; 5-hydroxytryptamine receptor 2A; serotonin 5-HT-2A receptor; serotonin 5HT-2 receptor; Serotonin receptor 2A
Gene Aliases: 5-HT2A; 5Ht-2; HTR2; HTR2A
UniProt ID: (Human) P28223, (Rat) P14842
Entrez Gene ID: (Human) 3356, (Rat) 29595
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.