Invitrogen
Manufacturer Code:PA572279
Catalog # PIPA572279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 223-258 amino acids from human HTR1E |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1E; 5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled; 5-hydroxytryptamine receptor 1E; HTR1E 5-HT1E 5-HT1e receptor 5-HT-1E 5HT1E Receptor HT1e receptor S31 Serotonin 5-HT-1e receptor Serotonin 1e receptor Serotonin receptor 1E; S31; Serotonin receptor 1E
Gene Aliases: 5-HT1E; HTR1E
UniProt ID: (Human) P28566
Entrez Gene ID: (Human) 3354
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.