Invitrogen
Manufacturer Code:PA5101281
Catalog # PIPA5101281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human 5-HT-1E corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1E; 5-HT1E 5-HT-1E 5HT1E Receptor 5-HT1e receptor 5-hydroxytryptamine (serotonin) receptor 1E G protein-coupled 5-hydroxytryptamine receptor 1E HT1e receptor HTR1E S31 Serotonin receptor 1E; 5-hydroxytryptamine (serotonin) receptor 1E, G protein-coupled; 5-hydroxytryptamine receptor 1E; S31; Serotonin receptor 1E
Gene Aliases: 5-HT1E; HTR1E
UniProt ID: (Human) P28566
Entrez Gene ID: (Human) 3354
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.