Invitrogen
Manufacturer Code:PA533292
Catalog # PIPA533292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 17 amino acid peptide from internal region of human 5HT1B Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1B; 5-HT-1D-beta; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; HTR1B 5-HT-1B 5-HT-1D-beta 5-HT1B 5-HT1b receptor 5-HT1DB 5HT1B Receptor HTR1DB Serotonin 5-HT-1b receptor Serotonin receptor 1B Serotonin 1b receptor HTR1D2 Serotonin 1D beta receptor; S12; Serotonin 1D beta receptor; Serotonin receptor 1B
Gene Aliases: 5-HT-1B; 5-HT-1D-beta; 5-HT1B; 5-HT1DB; HTR1B; HTR1D2; HTR1DB; S12
UniProt ID: (Human) P28222
Entrez Gene ID: (Human) 3351
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.