Invitrogen
Manufacturer Code:PA141069
Catalog # 01-676-664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to residues 8-26 and 263-278 of rat 5-HT1BR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1B; 5-HT-1D-beta; 5-HT1B; 5-HT1B receptor; 5-HT1B serotonin receptor; 5-HT<1B> receptor; 5-hydroxytryptamine (serotonin ) receptor 1B; 5-hydroxytryptamine (serotonin) receptor 1B, G protein-coupled; 5-hydroxytryptamine receptor 1B; S12; serotonin (5-hydroxytryptamine (5HT)) receptor type 1B; Serotonin 1D beta receptor; Serotonin receptor 1B
Gene Aliases: 5-HT-1B; 5-HT-1D-beta; 5-HT1B; 5-HT1DB; 5ht1b; HTR1B; HTR1D2; HTR1DB; S12
UniProt ID: (Human) P28222, (Mouse) P28334, (Rat) P28564
Entrez Gene ID: (Human) 3351, (Mouse) 15551, (Rat) 25075
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.