Invitrogen
Manufacturer Code:PA595035
Catalog # PIPA595035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human 5HT1A Receptor different from the related rat and mouse sequences by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1A; 5-HT1A; 5-HT1A receptor; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; 5-hydroxytryptamine receptor 1A 5-HT-1A 5-HT1A G-21 Serotonin receptor 1A HTR1A ADRB2RL1 ADRBRL1; G-21; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; Serotonin receptor 1A
Gene Aliases: 5-HT-1A; 5-HT1A; 5HT1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; HTR1A; PFMCD; RAT5HT1A
UniProt ID: (Human) P08908, (Mouse) Q64264, (Rat) P19327
Entrez Gene ID: (Human) 3350, (Mouse) 15550, (Rat) 24473
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.