Invitrogen
Manufacturer Code:PA577745
Catalog # PIPA577745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide KKSLNGQPGSGDWRRC corresponding to amino acid residues 251-266 of rat 5-Hydroxytryptamine Receptor 1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1A; 5-HT1A; 5-HT1A receptor; 5-HT1A Serotonin Receptor 1A; 5-hydroxytryptamine receptor 1A; G-protein coupled receptor 18; Serotonin receptor 1A
Gene Aliases: 5HT1A; Gpcr18; Htr1a; RAT5HT1A
UniProt ID: (Mouse) Q64264, (Rat) P19327
Entrez Gene ID: (Mouse) 15550, (Rat) 24473
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.