Invitrogen
Manufacturer Code:PA553716
Catalog # PIPA553716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HTR1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-HT-1A; 5-HT1A 5HT1a ADRB2RL1 ADRBRL1 5-HT-1A 5-HT1a receptor 5-hydroxytryptamine receptor 1A G protein coupled receptor G-21 guanine nucleotide-binding regulatory protein-coupled receptor serotonin receptor 1A; 5-HT1a receptor; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; G-21; guanine nucleotide-binding regulatory protein-coupled receptor; Serotonin receptor 1A
Gene Aliases: 5-HT-1A; 5-HT1A; 5HT1a; ADRB2RL1; ADRBRL1; G-21; HTR1A; PFMCD
UniProt ID: (Human) P08908
Entrez Gene ID: (Human) 3350
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.