Abnova
Manufacturer Code:PAB19759
Catalog # 89-121-286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide correspoding to internal region of mouse Htr1a. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Htr1a.
For Research Use Only
Protein Aliases: 5-HT-1A; 5-HT1A receptor; 5-hydroxytryptamine receptor 1A; G-protein coupled receptor 18; Gpcr18; Serotonin receptor 1A
Gene Aliases: Gpcr18; Htr1a
UniProt ID: (Mouse) Q64264
Entrez Gene ID: (Mouse) 15550
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.