Novus Biologicals
Manufacturer Code:NBP118694
Catalog # NBP118694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from Nter domain of Arabidopsis thaliana (Mouse-ear cress) HTR12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CENH3 Centromeric histone CENH3 F6F3.17 F6F3_17 Histone H3 like centromeric protein HTR12 HTR 12; Centromeric histone CENH3; Histone H3-like centromeric protein HTR12
Gene Aliases: AT1G01370; CENH3; CENTROMERIC HISTONE H3; F6F3.17; F6F3_17; HTR12
UniProt ID: (Arabidopsis) Q8RVQ9
Entrez Gene ID: (Arabidopsis) 839104
Molecular Function:
DNA binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.