Abnova
Manufacturer Code:MAB20179
Catalog # 89-336-579
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | IOG-8 |
Immunogen | A synthetic peptide corresponding to human HSPH1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: Antigen NY-CO-25; DKFZp686M05240 HSP105 HSP105A HSP105B KIAA0201 NY-CO-25; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa/110kDa protein 1; Heat shock 110 kDa protein; Heat shock protein 105 kDa
Gene Aliases: HSP105; HSP105A; HSP105B; HSP110; HSPH1; KIAA0201; NY-CO-25
UniProt ID: (Human) Q92598
Entrez Gene ID: (Human) 10808
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.