Abnova
Manufacturer Code:PAB8773
Catalog # 89-133-926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Hsph1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Hsph1.
For Research Use Only
Protein Aliases: 105kDa; 42 degrees C-HSP; AI790491; Heat shock 110 kDa protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein, 105 kDa; Heat shock-related 100 kDa protein E7I; HSP-E7I; Hsp105; HSP105 42 C-HSP; Hsp110
Gene Aliases: 105kDa; AI790491; hsp-E7I; Hsp105; Hsp110; hsp110/105; Hsph1; Kiaa0201
UniProt ID: (Mouse) Q61699
Entrez Gene ID: (Mouse) 15505
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.