Abnova
Manufacturer Code:PAB27072
Catalog # 89-128-722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human HSPH1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of HSPH1.
For Research Use Only
Protein Aliases: Antigen NY-CO-25; DKFZp686M05240; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa/110kDa protein 1; Heat shock 110 kDa protein; Heat shock protein 105 kDa; HSP105; HSP105A; HSP105B; KIAA0201; NY-CO-25
Gene Aliases: HSP105; HSP105A; HSP105B; HSP110; HSPH1; KIAA0201; NY-CO-25
UniProt ID: (Human) Q92598
Entrez Gene ID: (Human) 10808
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.