OriGene
Manufacturer Code:TA800847
Catalog # 50-167-8589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G10 |
Immunogen | Full length human recombit protein of human HSPD1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 60 kDa chaperonin; 60 kDa heat shock protein, mitochondrial; Chaperonin 60; CPN60; CPN60 GROEL HLD4 HSP-60 HSP60 HSP65 HuCHA60 SPG13; heat shock 60kDa protein 1 (chaperonin); Heat shock protein 60; heat shock protein 65; HSP-60; HuCHA60; Mitochondrial matrix protein P1; P60 lymphocyte protein; short heat shock protein 60 Hsp60s1
Gene Aliases: CPN60; GROEL; HLD4; HSP-60; HSP60; HSP65; HSPD1; HuCHA60; SPG13
UniProt ID: (Human) P10809
Entrez Gene ID: (Human) 3329
Molecular Function:
chaperone
chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.