Invitrogen
Manufacturer Code:PA569144
Catalog # PIPA569144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse HSPB7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiovascular heat shock protein; CVHSP; heat shock 27kD protein family member 7; heat shock 27kD protein family, member 7 (cardiovascular); heat shock 27kDa protein family member 7 (cardiovascular); heat shock 27kDa protein family, member 7 (cardiovascular); Heat shock protein 25 kDa 2; heat shock protein 25 kDa 2 (cardiovascular); Heat shock protein beta-7; heat shock protein family, alpha-crystallin-related, b7; heat shock protein family, member 7 (cardiovascular); HspB7; Protein p19/6.8
Gene Aliases: 27kDa; CVHSP; Hsp25-2; HSPB7; zgc:103426
UniProt ID: (Human) Q9UBY9, (Mouse) P35385
Entrez Gene ID: (Dog) 487423, (Horse) 100053742, (Human) 27129, (Rabbit) 100347539, (Rat) 50565, (Guinea pig) 100731021, (Mouse) 29818, (Zebrafish) 450019
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.