Novus Biologicals
Manufacturer Code:NBP216881
Catalog # NBP216881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human HSC70. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: constitutive heat shock protein 70; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; Heat shock 70 kDa protein 8; Heat shock 70 kDa protein 8 heat shock 70kD protein 8 heat shock 70kDa protein 8 heat shock cognate 71 kDa protein heat shock cognate protein 54 heat shock cognate protein 71-kDa HSC54 HSC70heat shock 70kd protein 10 HSC71 HSP71 HSP73constitutive heat shock protein 70 HSPA10 LAP1 lipopolysaccharide-associated protein 1 LPS-associated protein 1 MGC131511 MGC29929 NIP71 N-myristoyltransferase inhibitor protein 71; heat shock 70kd protein 10; heat shock 70kDa protein 8; Heat shock cognate 71 kDa protein; heat shock cognate protein 54; LAP-1; Lipopolysaccharide-associated protein 1; LPS-associated protein 1; N-myristoyltransferase inhibitor protein 71
Gene Aliases: HEL-33; HEL-S-72p; HSC54; HSC70; HSC71; HSP71; HSP73; HSPA10; HSPA8; LAP-1; LAP1; NIP71
UniProt ID: (Human) P11142
Entrez Gene ID: (Human) 3312
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.