Invitrogen
Manufacturer Code:PA517053
Catalog # PIPA517053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human HSPA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG-2 HS24/P52 HSPH2 RY hsp70 hsp70RY heat shock 70 kDa protein 4 heat shock 70-related protein APG-2 heat shock 70kD protein 4 heat shock protein 110 kDa hsp70 RY; epididymis secretory sperm binding protein Li 5a; Heat shock 70 kDa protein 4; Heat shock 70-related protein APG-2; heat shock 70kD protein 4; heat shock 70kDa protein 4; heat shock protein, 110 kDa; hsp70 RY; HSP70RY; Ischemia responsive 94 kDa protein
Gene Aliases: 70kDa; AI317151; APG-2; APG2; HEL-S-5a; HS24/P52; Hsp110; Hsp70; Hsp70RY; HSPA4; HSPH2; Irp94; mKIAA4025; RY
UniProt ID: (Human) Q9BUK9, (Rat) O88600
Entrez Gene ID: (Human) 3308, (Mouse) 15525, (Rat) 266759
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.