Invitrogen
Manufacturer Code:PA544330
Catalog # PIPA544330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human HSPA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 70 kDa heat shock protein; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock 70kDa protein 2 HSP70-2 HSP70-3; Heat shock protein 70.2; heat shock protein alpha 2; heat shock protein, 70 kDa 2; Heat shock-related 70 kDa protein 2; HST; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70
Gene Aliases: 70kDa; Hcp70.2; HSP70-2; HSP70-3; HSP70.2; HSP70.3; HSP70A2; HSPA2; Hspt70; Hst70
UniProt ID: (Human) P54652, (Rat) P14659, (Mouse) P17156
Entrez Gene ID: (Human) 3306, (Dog) 480355, (Goat) 102186695, (Pig) 100621324, (Rabbit) 100352959, (Rat) 60460, (Mouse) 15512
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.