OriGene
Manufacturer Code:TA500770
Catalog # 50-167-1913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F3 |
Immunogen | Full length human recombit protein of human HSPA1A produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein Li 5a; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 4; Heat shock 70-related protein APG-2; heat shock 70kD protein 1A; heat shock 70kD protein 4; heat shock 70kDa protein 1A; heat shock 70kDa protein 4; heat shock protein Apg-2; heat shock protein, 110 kDa; heat shock-induced protein; HEL-S-103 HSP70-1 HSP70-1A HSP70.1 HSP70I HSP72 HSPA1; hsp70 RY; HSP70-1; HSP70-1/HSP70-2; HSP70-2; HSP70.1/HSP70.2; HSP70RY
Gene Aliases: APG-2; APG2; HEL-S-103; HEL-S-5a; HS24/P52; Hsp70; HSP70-1; HSP70-1A; HSP70.1; HSP70I; hsp70RY; HSP72; HSPA1; HSPA1A; HSPA1B; HSPA4; HSPH2; HSX70; RY
UniProt ID: (Dog) Q7YQC6, (Human) P0DMV8, (Dog) Q2TFN9, (Human) Q9BUK9
Entrez Gene ID: (Dog) 403612, (Human) 3303, (Dog) 474680, (Human) 3308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.