Invitrogen
Manufacturer Code:MA538023
Catalog # PIMA538023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2351 |
Immunogen | A synthesized peptide derived from human HSPA12A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1700063D12Rik AI118035 AI840429 AW048913 AW556406 D5Mgi40 Heat shock 70 kDa protein 12A heat shock 70kD protein 12A heat shock 70kDa protein 12A Heat shock protein heat shock protein 12A heat shock protein family A (Hsp70) member 12A HSP HSPA12A KIAA0417 mKIAA0417; Heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A
Gene Aliases: 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; HSPA12A; KIAA0417; mKIAA0417
UniProt ID: (Human) O43301, (Mouse) Q8K0U4
Entrez Gene ID: (Human) 259217, (Rat) 307997, (Mouse) 73442
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.