Invitrogen
Manufacturer Code:PA526768
Catalog # PIPA526768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 398-427 amino acids from the central region of human HSPA12A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Heat shock 70 kDa protein 12A; heat shock 70 kDa protein 12A heat shock 70kD protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A
Gene Aliases: 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; HSPA12A; KIAA0417; mKIAA0417
UniProt ID: (Human) O43301, (Mouse) Q8K0U4
Entrez Gene ID: (Human) 259217, (Mouse) 73442
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.