Novus Biologicals
Manufacturer Code:NBP194014
Catalog # NBP194014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KLLVKDGTRWCTDVFDKFISADQSVALGELVKRSYTPAKPSQLVIVINIYSSEHDNVSFITDPGVKKCGTLRLDLTGTSGTAVPARREIQTLMQFGDTEIKATAIDIATSKSVKVGID |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ13874 heat shock 70 kDa protein 12A heat shock 70kD protein 12A heat shock 70kDa protein 12A KIAA0417; Heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A
Gene Aliases: HSPA12A; KIAA0417
UniProt ID: (Human) O43301
Entrez Gene ID: (Human) 259217
Molecular Function: Hsp70 family chaperone chaperone kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.