Novus Biologicals
Manufacturer Code:NBP267395
Catalog # NBP267395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SY46-01 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Heat shock 84 kDa; Heat shock 84 kDa heat shock 90kD protein 1 beta heat shock 90kDa protein 1 beta heat shock protein 90kDa alpha (cytosolic) class B member 1 heat shock protein beta heat shock protein HSP 90-beta HSP 84 HSP84 HSP90-BETA HSP90BHSP 90 HSPC2D6S182 HSPCBFLJ26984; heat shock 90kD protein 1, beta; heat shock protein 90 kDa; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; HSP 84; HSP 90; HSP90-beta
Gene Aliases: D6S182; HSP84; HSP90AB1; HSP90B; HSPC2; HSPCB
UniProt ID: (Human) P08238
Entrez Gene ID: (Human) 3326
Molecular Function:
Hsp90 family chaperone
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.