Novus Biologicals
Manufacturer Code:NB6001469
Catalog # NB6001469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Gel Shift (GS) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A3 |
Immunogen | Human recombinant HSP70 overexpressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: dnaK-type molecular chaperone HSP70-1; dnaK-type molecular chaperone HSP70-1 FLJ54303 FLJ54370 FLJ54392 FLJ54408 FLJ75127 Heat shock 70 kDa protein 1/2 heat shock 70 kDa protein 1A/1B heat shock 70kD protein 1A heat shock 70kDa protein 1A heat shock-induced protein HSP70.1/HSP70.2 HSP70-1 HSP70-1/HSP70-2 HSP70-1A HSP70I HSP72 HSPA1 HSPA1B; epididymis secretory protein Li 103; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1A; heat shock 70kDa protein 1A; heat shock-induced protein; HSP70-1; HSP70-1/HSP70-2; HSP70-2; HSP70.1/HSP70.2
Gene Aliases: HEL-S-103; HSP70-1; HSP70-1A; HSP70.1; HSP70I; HSP72; HSPA1; HSPA1A; HSPA1B; HSX70
UniProt ID: (Human) P0DMV8
Entrez Gene ID: (Human) 3303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.