Proteintech
Manufacturer Code:109951AP150UL
Catalog # 50-556-618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HSP70 Fusion Protein Ag1446 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 68 kDa heat shock protein; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1/2 heat shock 70kDa protein 1A HSP70 HSP70 1 HSP70 1/HSP70 2 HSP70 1A HSP70.1/HSP70.2 HSP70I HSP72 HSPA1 HSPA1A HSPA1B; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 1A; heat shock 70kDa protein 1A; heat shock protein 70-1; heat shock protein, 70 kDa 3; heat shock-induced protein; Hsp68; HSP70-1; HSP70-1/HSP70-2; HSP70-2; HSP70.1/2; HSP70.1/HSP70.2; HSP70.3; inducible heat shock protein 70
Gene Aliases: HEL-S-103; hsp68; HSP70-1; HSP70-1A; Hsp70-2; Hsp70-3; HSP70.1; Hsp70.3; Hsp70A1; HSP70I; HSP72; HSPA1; HSPA1A; HSPA1B; Hspa2; HSX70
UniProt ID: (Human) P0DMV8, (Mouse) Q61696, (Rat) P0DMW1
Entrez Gene ID: (Human) 3303, (Mouse) 193740, (Rat) 24472
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.